Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Dropping Bottles

LDPE Dropping Bottles, Capacity: 60 mL, 60 ml (2 oz.), 18 mm
  470123-918
 :   Qorpak
 :  501186
Dropping Bottles
  470123-918
 :  Qorpak
Dropping Bottles
Dropping Bottles

LDPE Dropping Bottles, Capacity: 60 mL, 60 ml (2 oz.), 18 mm

 
$90.05
   
Pack of 12
 

Dropping bottles are made of low-density polyethylene.

  • White Polypropylene Dropper and Nasal Cap
  • Molded Plastic Screw Cap Seals at Tip
  • Low-Density Polyethylene

They feature a separable dropping tip.

The molded plastic screw cap seals at the tip.

 
  • Cap Size:
    18 mm
  • Capacity:
    60 ml (2 oz.)
  • Description:
    LDPE Dropping Bottles, Capacity: 60 mL