Orders LinkContactUsLinkComponent
Levitating Magnets Set
Levitating Magnets Set
Levitating Magnets Set
Catalog # 470122-658
Supplier:   DOWLING MAGNETS,LLC
Supplier #:  SS10
Levitating Magnets Set
Catalog # 470122-658
Levitating Magnets Set
Levitating Magnets Set
Levitating Magnets Set
Price
$27.55
Log in for availability
Kit of 1
Product Details & Documents
Floating Magnets Seem to Defy Gravity
  • Includes five brightly colored ceramic ring magnets each 28mm in diameter and a 15cm wooden dowel on which to mount them
Bring a little levitation to your classroom with these ring magnets. They graphically demonstrate both attraction and repulsion of magnetism. Replacement magnet available.
Specifications
  • Description:
    Levitating Magnets Set