ADC® Macintosh Blades
Macintosh Blades, Child
Catalog # 470119-686
Supplier: AMERICAN DIAGNOSTIC CORP CP
: 4072
Jump to:
Product Details & Documents
Stainless steel construction, satin, non-glare finish.
This curved blade designs is fully compatible with all standard hook-on handles.
: Conform to ASTM (F965) and ISO (7376-1).
Specifications
- Description:Macintosh Blades, Child