Orders LinkContactUsLinkComponent
Simulaids® Birthing Simulator
Simulaids® Birthing Simulator
Replacement Umbilical Cords (Pk/12)
Catalog # 470119-262
Supplier:   Nasco
Supplier #:  110-159
Simulaids® Birthing Simulator
Catalog # 470119-262
Supplier:  Nasco
Simulaids® Birthing Simulator
Simulaids® Birthing Simulator
Replacement Umbilical Cords (Pk/12)
Price
$128.25
Log in for availability
Each
Product Details & Documents
Practice Through All Stages Of The Birthing Process
  • Belly overlay contains a fetus in fluid and can be palpated for position
  • Lifelike pelvic cavity
  • Practice multiple procedures
  • Includes a soft bag for storage and transport
An anatomically correct pelvic model with full term newborn and placenta to give your trainees realistic practice in multiple techniques, and to learn the procedure for emergency childbirth. Disposable umbilical cords with clamps, easily replaceable extra vulva, powder to make simulated blood, a modular pregnant belly overlay with permanently installed fetus for training practice of the Leopold’s maneuver to determine the fetus’ lie by palpating its skull and kneecaps, and a clear abdominal overlay to see positioning are all included. The lifelike pelvic cavity has pronounced pelvic landmarks, located spinal column, angled birth canal, ileum, ischium, sacrum, sacro spinious ligaments, and greater sciatic notch. The unit is capable of demonstrating shoulder distocia, fetal attitude, fetal station, fetal lie, cord prolapse, compound presentation, and cesarean section incision locations can be discussed. The fetus provided is of a realistic newborn size with fontanels and cranial sutures.
Specifications
  • Description:
    Replacement Umbilical Cords (Pk/12)