Order Entry
Canada
Orders LinkContactUsLinkComponent
DC Motor Kit, Single Kit

DC Motor Kit, Single Kit

DC Motor Kit, Class Set of 12 Kits, 12 Lab groups
Catalog # 470111-204
Supplier:   Wards
Supplier #:  DCMKT-12
DC Motor Kit, Single Kit
Catalog # 470111-204
Supplier:  Wards
DC Motor Kit, Single Kit
DC Motor Kit, Single Kit

DC Motor Kit, Class Set of 12 Kits, 12 Lab groups

Price
$194.25
Log in for availability
Pack of 12
Product Details & Documents

This kit is a learn-by-building introduction to a motor’s basic components.

  • Focus on Poles, Armatures, and Switches
  • Class Set of 12 or Individual Kits Available
  • One AA Battery Required

Students gain insight into field poles, armatures, and switches.

The kit requires one AA battery, sold separately. It's available individually or as an economical class set of 12.

Ordering information: This kit assembly instructions, a 9 x 6.5 cm mounting base, two coils of insulated wire (200 cm long), a field pole, a mounting bracket, two armature halves, one piece of insulating tubing (4 cm long), a motor shaft (6.5 cm long), two commutator insulators, two bronze wire brushes, two shaft supports, four paper fasteners, a AA battery holder, six screws, and six nuts.

Specifications
  • For Use With:
    12 Lab groups
  • Description:
    DC Motor Kit, Class Set of 12 Kits
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.