Orders LinkContactUsLinkComponent
Blocks and Refraction Plates
Blocks and Refraction Plates
Blocks and Refraction Plates, Acrylic Block, 70 x 70 x 9mm
Catalog # 470110-648
Supplier:   Wards
Supplier #:  PH0568E
Blocks and Refraction Plates
Catalog # 470110-648
Supplier:  Wards
Blocks and Refraction Plates
Blocks and Refraction Plates
Blocks and Refraction Plates, Acrylic Block, 70 x 70 x 9mm
Price
19,85 $
Log in for availability
Each
Product Details & Documents
Parallel-Sided Blocks for Ray Optics
Ideal for refraction and internal reflection studies as well as index of refraction measurements, these clear blocks are free of bubbles and streaks and have polished surfaces. Acrylic blocks and refraction plates have frosted ends, glass block is completely clear.
Specifications
  • Description:
    Blocks and Refraction Plates, Acrylic Block, 70 x 70 x 9mm