Equilateral Prisms
Equilateral Prisms, Acrylic, 25 x 12 mm
Catalog # 470110-640
Supplier: Wards
Supplier #: PH0554A
Jump to:
Product Details & Documents
For all Optical Applications
- These prisms are polished on all faces
- They are made of shatter-resistant acrylic material or optical quality glass
These prisms are polished on all faces. They are made of shatter-resistant acrylic material or optical quality glass. The length of the sides and the height are given for each prism.
Specifications
- Description:Equilateral Prisms, Acrylic, 25 x 12 mm