Order Entry
Canada
Orders LinkContactUsLinkComponent
Triangular Supports with Rods

Triangular Supports with Rods

4", ⅜", 20"
Catalog # 470105-200
Supplier #:  TSR420
Triangular Supports with Rods
Catalog # 470105-200
Triangular Supports with Rods
Triangular Supports with Rods

4", ⅜", 20"

Price
$54.65
Log in for availability
Each
Product Details & Documents

Heavy cast-iron triangular bases.

  • Low flat legs on triangular supports provide excellent stability

Black enamel finish is extremely resistant to wear.

Delivery information: Includes a removable, zinc-plated steel rod.

Specifications
  • Rod length:
    20"
  • Base dimensions:
    4"
  • Rod diameter:
    ⅜"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.