Orders LinkContactUsLinkComponent
LaMotte® Secchi Disk
LaMotte® Secchi Disk
Secchi Disk Only, 8"
Catalog # 470101-430
Supplier:   LaMotte
Supplier #:  0171
LaMotte® Secchi Disk
Catalog # 470101-430
Supplier:  LaMotte
LaMotte® Secchi Disk
LaMotte® Secchi Disk
Secchi Disk Only, 8"
Price
$74.00
Log in for availability
Each
Product Details & Documents
This 8", black and white disk is used to determine the turbidity of natural water.
  • Black and White design
  • Sounding lead and line included
  • Function: to determine the turbidity of water
  • Aligned with GLOBE protocols
The disk is raised or lowered several times to determine the average depth at which it disappears from sight.
Ordering information: 470112-626 is complete set, containing a secchi disk and a calibrate line. 470004-364 is the standalone rope, and 470101-430 the standalone disk.
Specifications
  • Description:
    Secchi Disk Only
  • O.D.:
    8"