$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
CD EXPLORING THE ROCK CYCLE MAC/WIN SNGL
470038-332
:
MERCURY PRINT PRODUCTIONS SE
CD EXPLORING THE ROCK CYCLE MAC/WIN SNGL
470038-332
:
MERCURY PRINT PRODUCTIONS SE
:
6801020
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International