Orders LinkContactUsLinkComponent
Ward's® Plastic Specimen Trays
Ward's® Plastic Specimen Trays
WARD’S Plastic Specimen Trays, 12-Compartment Tray
Catalog # 470036-900
Supplier #:  B19180/13-SP
Ward's® Plastic Specimen Trays
Catalog # 470036-900
Ward's® Plastic Specimen Trays
Ward's® Plastic Specimen Trays
WARD’S Plastic Specimen Trays, 12-Compartment Tray
Price
$9.30
Log in for availability
Each Case of 380
Product Details & Documents
Keep Your Collections Organized
Use our vacuum-formed compartmented trays alone to keep small items separate, or in our collection boxes for permanent dustfree storage of your samples. The white nonglare finish will not yellow with age.
12-Compartment Tray
Tray size: 6 3/4"L x 6 3/4"W x 5/8"H; cavity size: 2"L x 1 1/2"W.
16-Compartment Tray
Tray size: 7"L x 7"W x 5/8"H; cavity size: 1 1/2"L x 1 1/2"W.
24-Compartment Tray
Tray size: 12 1/2"L x 9 1/4"W x 5/8"H; cavity size: 2 1/4"L x 1 7/8"W.
36-Compartment Tray
Tray size: 12 1/2"L x 9 1/4"W x 5/8"H; cavity size: 2"L x 1 1/2"W.
45-Compartment Tray
Tray size: 16"L x 12 1/2"W x 5/8"H; cavity size: 2 1/2"L x 1 3/4"W.
50-Compartment Tray
Tray size: 15 3/4"L x 12 1/2"W x 3/4"H; cavity size: 2 1/4"L x 1 3/8"W.
Specifications
  • Description:
    WARD’S Plastic Specimen Trays, 12-Compartment Tray