Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VANADINITE XLS ON 2X3 MATRIX ARIZONA
Catalog # 470026-696
Supplier:
ADAM MOORE MO
Expand image
VANADINITE XLS ON 2X3 MATRIX ARIZONA
Catalog # 470026-696
Supplier:
ADAM MOORE MO
Supplier Number:
TUCSON0085
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH