$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
PYRITE SGLE XL 1-3CM PENT/DODEC/CUBE ES
470026-460
:
ADAM MOORE MO
PYRITE SGLE XL 1-3CM PENT/DODEC/CUBE ES
470026-460
:
ADAM MOORE MO
:
TUCSON0080
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH