Order Entry
Canada
Orders LinkContactUsLinkComponent
Lab Label Roll

Lab Label Roll

Lab label roll, ¾", 100"
Catalog # 470019-788
Supplier:   ROLL PRODUCTS,INC.
Supplier #:  15-1970
Lab Label Roll
Catalog # 470019-788
Supplier:  ROLL PRODUCTS,INC.
Lab Label Roll
Lab Label Roll

Lab label roll, ¾", 100"

Price
$27.05
Log in for availability
Each
Product Details & Documents

Better than gummed labels, these pressure-sensitive labels adhere securely, yet can be easily removed.

  • Autoclavable to 260 °C

The labels are on a continuous roll.

Specifications
  • Description:
    Lab label roll
  • Length:
    100"
  • Width:
    ¾"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.