Vertical School Seismometer
Product Details & Documents
Simple Yet Sensitive Instrument.
- Detect earthquakes before the media reports them
- Includes software to analyze data
- Dust cover protects seisomometer and damping oil from dust and drafts (sold separately)
With the Vertical School Seismometer, students can watch a developing event unfold and analyze the recordings of generated waves.
Built on the proven Incorporated Research Institutions for Seismology standards, this instrument can be placed in any stable, relatively undisturbed location, and it will deliver P and S-wave earthquake signals all day.
The seisomometer comes with Windows software which displays a full day’s recordings on a single screen, automatically saves, and organizes the data for later analysis.
The software supports identification of different types of ground waves in an earthquake, extraction of the earthquake signal, and determination of the distance and intensity of the quake.
System Requirements: Windows 2000 or higher, USB
Specifications
- Description:Seismometer Dust Cover
- Height:16"
- Length:23"
- Width:12"
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



