Stake Flags
Stake Flags, Orange
Catalog # 470018-560
Supplier: Peco Sales
Supplier #: 39549
Jump to:
Product Details & Documents
A versatile marking tool.
- Aligned with GLOBE protocols
Use these flags attached to steel wire stakes to mark areas for field or watershed studies, or use them for other activities, such as species identification. The flags are solid-colored, colorfast vinyl and flexible. They are also mold and mildew resistant. The flags are 4" x 5", the stakes are 30" high, and they come in packages of 100.
Specifications
- Description:Stake Flags, Orange