Orders LinkContactUsLinkComponent
Increment Borers
Increment Borers
Increment Borer, 10" Bore Length
Catalog # 470018-550
Supplier:   Peco Sales
Supplier #:  63082
Increment Borers
Catalog # 470018-550
Supplier:  Peco Sales
Increment Borers
Increment Borers
Increment Borer, 10" Bore Length
Price
$629.45
Log in for availability
Each
Product Details & Documents
Used to Examine Wood Quality and Measure Growth Rates
These borers are made of the finest steel and coated with Teflon to reduce friction heat, which is an important consideration when working with growing trees. The Teflon, in combination with the three-thread style, also makes the borers easy to extract.
Specifications
  • Description:
    Increment Borer, 10" Bore Length