Order Entry
Canada
Orders LinkContactUsLinkComponent
Solid Balls for Physics Experiments

Solid Balls for Physics Experiments

Solid Steel Ball, 25 mm, 1"
Catalog # 470016-974
Supplier #:  CARBONGR100 25
Solid Balls for Physics Experiments
Catalog # 470016-974
Solid Balls for Physics Experiments
Solid Balls for Physics Experiments

Solid Steel Ball, 25 mm, 1"

Price
$11.70
Log in for availability
Each
Product Details & Documents

Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.

Caution: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.

Specifications
  • Description:
    Solid Steel Ball, 25 mm
  • Diameter:
    1"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.