Orders LinkContactUsLinkComponent
Dropping Bottles
Dropping Bottles
30 ml (1 oz.), —
Catalog # 470016-200
Supplier:   Qorpak
Supplier #:  501182
Dropping Bottles
Catalog # 470016-200
Supplier:  Qorpak
Dropping Bottles
Dropping Bottles
30 ml (1 oz.), —
Price
$109.50
Log in for availability
Pack of 12
Product Details & Documents
Dropping bottles are made of low-density polyethylene.
  • White Polypropylene Dropper and Nasal Cap
  • Molded Plastic Screw Cap Seals at Tip
  • Low-Density Polyethylene
They feature a separable dropping tip.
The molded plastic screw cap seals at the tip.
Documents
Specifications
  • Cap Size:
  • Capacity:
    30 ml (1 oz.)