Miscellaneous Convenience Items
Spring, Compressible 3 cm
Catalog # 470012-744
Supplier: STANLEY SPRING & STAMPING
: 2804500
Jump to:
Product Details & Documents
Caution: WARNING: CHOKING HAZARD. Not for children under 3 years.
Specifications
- Description:Spring, Compressible 3 cm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Funnel, Poly, 114 mL (4 oz)
Product Information
- Catalog #AvailabilityPrice
- Single sheave
Product Information
- Catalog #AvailabilityPrice
- Ball-Bearing Clamp Pulley
Product Information
- Catalog #AvailabilityPrice
- Battery Holder, 9 V
Product Information
- Catalog #AvailabilityPrice
- Weight, Hooked, 10 g to 1000 g, Cast Iron, Set/9Weight, Hooked, 10 g to 1000 g, Cast Iron, Set/9470003-584Log in for availabilityKit of 1 - $130.05
Product Information
- Catalog #ColorLengthAvailabilityPrice
- Braided Nylon pulley cord, tensile strength 200 lbs. testBraided Nylon pulley cord, tensile strength 200 lbs. test470005-414White9 m (10 yds.)Log in for availabilityEach - $31.00
Product Information
- Catalog #AvailabilityPrice
- Assorted Plastic Pulleys
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



