Hook Collars
Hook Collar, Fits 13 mm (0.5") Rod
Catalog # 470006-700
Supplier: United Scientific Supplies
Supplier #: HKCL01
Jump to:
Product Details & Documents
Quickly add a hook to a support rod.
- Plated steel
- Thumbscrew
Plated steel collar clamps feature thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
Specifications
- Description:Hook Collar, Fits 13 mm (0.5") Rod
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



