Order Entry
Canada
Orders LinkContactUsLinkComponent
DC Motor Kit, Single Kit

DC Motor Kit, Single Kit

DC Motor Kit, Single Kit, 1 Lab group
Catalog # 470003-836
Supplier:   Wards
Supplier #:  PH1229
DC Motor Kit, Single Kit
Catalog # 470003-836
Supplier:  Wards
DC Motor Kit, Single Kit
DC Motor Kit, Single Kit

DC Motor Kit, Single Kit, 1 Lab group

Price
$22.95
Log in for availability
Kit of 1
Product Details & Documents

This kit is a learn-by-building introduction to a motor’s basic components.

  • Focus on Poles, Armatures, and Switches
  • Class Set of 12 or Individual Kits Available
  • One AA Battery Required

Students gain insight into field poles, armatures, and switches.

The kit requires one AA battery, sold separately. It's available individually or as an economical class set of 12.

 : This kit assembly instructions, a 9 x 6.5 cm mounting base, two coils of insulated wire (200 cm long), a field pole, a mounting bracket, two armature halves, one piece of insulating tubing (4 cm long), a motor shaft (6.5 cm long), two commutator insulators, two bronze wire brushes, two shaft supports, four paper fasteners, a AA battery holder, six screws, and six nuts.

Specifications
  • For Use With:
    1 Lab group
  • Description:
    DC Motor Kit, Single Kit