Disposable Impression Trays
Disposable Impression Trays, Lower, 12 Trays
Catalog # 470020-462
Supplier: FRANKLIN DENTAL SUPPLY
Supplier #: 1004
Jump to:
Product Details & Documents
Inexpensive Recreations for Labwork
- Autoclavable
- Easy to Clean
Cast impressions of teeth to identify placement and study bite marks for forensic odontology lessons. This set of disposable dental impression trays is autoclavable and available for upper and lower jaw impressions. Set
of 12.
Specifications
- Description:Disposable Impression Trays, Lower
- Includes:12 Trays