Thin Stem Pipettes
4 ml, 25
Catalog # 470019-808
Supplier: Kemtec
Supplier #: 4-424C
Jump to:
Product Details & Documents
Versatile pipettes provide consistent drop size.
- Chemically inert and non-toxic
- Can be stretched to form micropipettes and other micro lab ware
These pipettes serve as ideal micro-reagent vials that can be heat-sealed and frozen.
Specifications
- Capacity:4 ml
- Drops per ml:25