Order Entry
Canada
Orders LinkContactUsLinkComponent
Calibration Buffer Solutions

Calibration Buffer Solutions

Calibration Buffer Solution, 1 pint, 4.01
Catalog # 470019-678
Supplier:   Antyila Scientific
Supplier #:  00654-00
Calibration Buffer Solutions
Catalog # 470019-678
Supplier:  Antyila Scientific
CAS Number:  
Calibration Buffer Solutions
Calibration Buffer Solutions

Calibration Buffer Solution, 1 pint, 4.01

CAS Number:  
Price
$34.40
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Always read the correct pH

  • Accurate Results
  • Easy clean up

Ready to use solutions to calibrate your pH meter.

Documents

Specifications

Specifications

  • Description:
    Calibration Buffer Solution
  • pH:
    4.01
  • Volume:
    1 pint

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.