Orders LinkContactUsLinkComponent
Pulleys
Pulley, Quadruple Sheave
  470005-698
 :   Wards
 :  PH0280F
Pulleys
  470005-698
 :  Wards
Pulleys
Pulleys
Pulley, Quadruple Sheave
$3.85
  31/12/2026
Each
These free-running plastic pulleys are durable in design.
  • Plastic Construction
  • Durable
  • Nickel-Plated Steel Blocks with Hooks
The sheaves are 50 mm in Ø and made of rigid plastic.
The nickel-plated steel blocks have hooks at the top and bottom.
  • Description:
    Pulley, Quadruple Sheave