Multi Purpose Fire Extinguishers
Multi-purpose fire extinguisher, 10 lbs., 4A60B:C
Supplier: Logistics Supply
: 466204K
Jump to:
Product Details & Documents
For use on Fire Types (Classes) A, B, and C.
- Pressure gauge provides at-a-glance status
- Easy-to-pull safety pin
- Powder coated cylinder for corrosion protection
- 6-year limited warranty
These ideal classroom fire extinguishers are effective against all three types of fire; wood and paper, liquids and grease, and electrical. Both extinguishers contain dry chemical powder, pressurized with carbon dioxide gas. The powder is discharged in a blanket-like stream at a range of more than four meters to smother the fire. Both extinguishers are supplied with wall-mounting hardware.
: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Spécifications
- Description:Multi-purpose fire extinguisher, 10 lbs.
- Ul rating:4A60B:C



