Orders LinkContactUsLinkComponent
Fingerprint Analysis Lab Activity | Avantor
Fingerprint Analysis Lab Activity
Fingerprint Analysis Kit Refill, 6
Catalog # 470002-518
Supplier:   Kemtec
Fournisseur #:  10-504R
Fingerprint Analysis Lab Activity
Catalog # 470002-518
Supplier:  Kemtec
Fingerprint Analysis Lab Activity
Fingerprint Analysis Lab Activity
Fingerprint Analysis Kit Refill, 6
Price
$181.25
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A fun and educational introduction to criminalistics.
  • Grades 7-14
  • Multiple methods for detecting latent prints
Your students will answer the question “Whodunit?” while learning how to dust and develop latent fingerprints. They “lift” sets of prints from various surfaces, turning them over for others to analyze. After comparing the prints to standards developed by the FBI, and working with “Modus Operandi” worksheets, the class will identify the suspects. The kit includes instructor’s and assessment guides, student data sheets, and all materials but the fingerprints for 24 students.
Spécifications
  • Description:
    Fingerprint Analysis Kit Refill
  • No. of lab groups:
    6
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.