Orders LinkContactUsLinkComponent
Turbidity
Turbidity
Turbidity Refill Kit
Catalog # 470002-188
Supplier:   LaMotte
Supplier #:  7520-H
Turbidity
Catalog # 470002-188
Supplier:  LaMotte
Turbidity
Turbidity
Turbidity Refill Kit
Price
$32.60
Log in for availability
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: Five to 200 JTU
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
A compact, waterproof carrying case contains reagents, labware, and step-by-step instructions.
The tests have a range of 5 to 200 JTU.
Specifications
  • Description:
    Turbidity Refill Kit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)