Orders LinkContactUsLinkComponent
Correlative Microscopy Coverslips, Electron Microscopy Sciences
Correlative Microscopy Coverslips, Electron Microscopy Sciences
Cover slips for correlative microscopy, grid 0.5 mm, 20×20 mm
Catalog # 102773-606
Supplier #:  66108-02
Correlative Microscopy Coverslips, Electron Microscopy Sciences
Catalog # 102773-606
Correlative Microscopy Coverslips, Electron Microscopy Sciences
Cover slips for correlative microscopy, grid 0.5 mm, 20×20 mm
Price
$324.73
Log in for availability
Each Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS introduces a simple coverslip with a grid image that allows cells to grow and later be analyzed under brightfield or fluorescence microscopy before being further processed and sectioned for EM or other analysis.
  • Reticle film has been thoroughly tested with cell culture
  • Good growth of cell culture (equivalent to conventional media)
  • Good adhesion to the substrate without cell polylysine
  • Resistant to normal chemicals used in electron microscopy
  • No oxygen retention, compatible resin LR White
  • Good optical quality in brightfield & UV fluorescence
  • Excellent transparency
  • Does not deform at temperatures (positive 100C and negative liquid N2)
  • Rigid, does not float in the middle of culture
  • Easy to handle and cut with a knife or micro-punch
  • Simple sterilisation using alcohol or UV
  • Detaches easily from resin after polymerisation
  • Low cost
Applications include Light Microscopy, Fluorescence, Scanning Electron Microscopy (SEM), Transmission Electron Microscopy (TEM), and High-Pressure Cryofixation.
Specifications
  • Description:
    Cover slips for correlative microscopy, grid 0.5 mm
  • L×W:
    20×20 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)