Order Entry
Canada
Orders LinkContactUsLinkComponent
Cutters, Carbide Insert, Excelta

Cutters, Carbide Insert, Excelta

Cutters, small taper head, hard wire, 2 star quality, carbide inserts, Stainless steel, 106.25 mm (4.25"), 12°
Catalog # 89411-862
Supplier:   Excelta
Supplier #:  2876EHT
Cutters, Carbide Insert, Excelta
Catalog # 89411-862
Supplier:  Excelta
Cutters, Carbide Insert, Excelta
Cutters, Carbide Insert, Excelta

Cutters, small taper head, hard wire, 2 star quality, carbide inserts, Stainless steel, 106.25 mm (4.25"), 12°

Price
$203.23
Log in for availability
Each
Product Details & Documents

This compound action cutter is ideal for cutting small hard wire.

  • Carbide insert cutting edges for tough hard wire
  • Compound-Action means less pressure on hands
  • Dual leaf springs for ease of use
  • Stainless steel shank for corrosion resistants

These multi-purpose medium round hard wire cutters feature stainless steel construction for corrosion resistance and carbide inserts for maximum blade life. Hard wire cutting capacity is 0.010 to 0.020". The grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq. Interestingly, the 149AT is a compound-action cutter, which requires much less force to operate.

Specifications
  • Description:
    Cutters, small taper head, hard wire, 2 star quality, carbide inserts
  • Material:
    Stainless steel
  • Curvature:
    12°
  • Overall length:
    106.25 mm (4.25")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.