Order Entry
Canada
Orders LinkContactUsLinkComponent
Scissors, Trimmer, Pattern Cutting, Excelta Corp®

Scissors, Trimmer, Pattern Cutting, Excelta Corp®

Scissors, trimmer, 2 star quality, 7" (175 mm), Stainless steel AISI 440, 825 mm (3¼"), Straight
Catalog # 89411-762
Supplier:   Excelta
Supplier #:  340
Scissors, Trimmer, Pattern Cutting, Excelta Corp®
Catalog # 89411-762
Supplier:  Excelta
Scissors, Trimmer, Pattern Cutting, Excelta Corp®
Scissors, Trimmer, Pattern Cutting, Excelta Corp®

Scissors, trimmer, 2 star quality, 7" (175 mm), Stainless steel AISI 440, 825 mm (3¼"), Straight

Price
$54.83
Log in for availability
Each
Product Details & Documents

These two star trimmer scissors are used for pattern cutting and general purpose use.

  • Ideal scissors for pattern cutting
  • Three different sizes to choose from

They are chrome-plated and come in various lengths.

Specifications
  • Description:
    Scissors, trimmer, 2 star quality
  • Length:
    7" (175 mm)
  • Material:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade length:
    825 mm (3¼")