Orders LinkContactUsLinkComponent
MQuant® pH Test Strips, Supelco®,MilliporeSigma
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Alkalit® pH-Indicator strips, non-bleeding, intermediate range, 100, 7.5-8.0-8.5-9.0-9.5-10.0-10.5-
11.0-11.5-12.0-12.5-13.0-13.5-14.0, 7.5 - 14, 0.5
Catalog # EM1.09532.0001
Supplier #:  1095320001
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Catalog # EM1.09532.0001
Supplier:  
MQuant® pH Test Strips, Supelco®,MilliporeSigma
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Alkalit® pH-Indicator strips, non-bleeding, intermediate range, 100, 7.5-8.0-8.5-9.0-9.5-10.0-10.5-
11.0-11.5-12.0-12.5-13.0-13.5-14.0, 7.5 - 14, 0.5
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BDH PH TEST STRIP INTER RANGE 7-14 PK5 Catalog #BDH35312.607
Product BDH35312.607 is a replacement for EM1.09532.0001 and has the same packaging and specifications, but may be a different quantity.
STRIP PH INDICATOR PH7.5-14 1SET Catalog #CA1.09532.0001
Product CA1.09532.0001 is a replacement for EM1.09532.0001 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
These strips provide a convenient method for accurate colorimetric pH indication.
The strips feature covalently bound indicator dyes so that they will not bleed even in strong alkaline solutions. Strips can be immersed in samples for extended periods, so that even weakly buffered solutions can be accurately tested without contaminating the sample. The universal strip tests the full 0–14 pH range with a sensitivity of 1 pH unit. Intermediate range indicators provide a sensitivity of 0.3–0.5 pH units and narrow range strips provide a sensitivity of 0.2–0.3 pH units.
MilliporeSigma # 1.09531.0001 replaces and is identical to MilliporeSigma # 9586-1.
MilliporeSigma # 1.09531.0007 replaces and is identical to MilliporeSigma # 9586-3.
MilliporeSigma # 1.09532.0001 replaces and is identical to MilliporeSigma # 9587-1.
MilliporeSigma # 1.09533.0001 replaces and is identical to MilliporeSigma # 9588-1.
MilliporeSigma # 1.09533.0007 replaces and is identical to MilliporeSigma # 9588-3.
MilliporeSigma # 1.09535.0001 replaces and is identical to MilliporeSigma # 9590-1.
MilliporeSigma # 1.09535.0007 replaces and is identical to MilliporeSigma # 9590-3.
MilliporeSigma # 1.09540.0001 replaces and is identical to MilliporeSigma # 9580-1.
MilliporeSigma # 1.09541.0001 replaces and is identical to MilliporeSigma # 9581-1.
MilliporeSigma # 1.09542.0001 replaces and is identical to MilliporeSigma # 9582-1.
MilliporeSigma # 1.09542.0007 replaces and is identical to MilliporeSigma # 9582-3.
MilliporeSigma # 1.09543.0001 replaces and is identical to MilliporeSigma # 9583-1.
MilliporeSigma # 1.09543.0007 replaces and is identical to MilliporeSigma # 9583-3.
MilliporeSigma # 1.09545.0001 replaces and is identical to MilliporeSigma # 9585-1.
MilliporeSigma # 1.09584.0001 replaces and is identical to MilliporeSigma # 9578-1.
Accessories information: Analyze your nitrate test strips with the reference card and with the free MQuant® StripScan app. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.The StripScan app allows the customer to analyze the MQuant® test strips with your smartphone for fast, convenient, and precise results which can be easily graphically presented and shared.
Specifications
  • Description:
    Alkalit® pH-Indicator strips, non-bleeding, intermediate range
  • pH range:
    7.5 - 14
  • pH graduation:
    7.5-8.0-8.5-9.0-9.5-10.0-10.5-
    11.0-11.5-12.0-12.5-13.0-13.5-14.0
  • Resolution:
    0.5
  • No. of strips:
    100