Orders LinkContactUsLinkComponent
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Acilit® pH-Indicator strips, non-bleeding, intermediate range, 600, 0-0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-
4.5-5.0-5.5-6.0, 0 - 6, 0.5
Catalog # EM1.09531.0007
 :  1095310007
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Catalog # EM1.09531.0007
Supplier:  
MQuant® pH Test Strips, Supelco®,MilliporeSigma
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Acilit® pH-Indicator strips, non-bleeding, intermediate range, 600, 0-0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-
4.5-5.0-5.5-6.0, 0 - 6, 0.5
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
COLORPHAST PH0-6 6X100/EA Catalog #CA1.09531.0007
BDH PH TEST STRIP INTER RANGE0-6 Catalog #BDH35310.601
Product Details & Documents
These strips provide a convenient method for accurate colorimetric pH indication.
The strips feature covalently bound indicator dyes so that they will not bleed even in strong alkaline solutions. Strips can be immersed in samples for extended periods, so that even weakly buffered solutions can be accurately tested without contaminating the sample. The universal strip tests the full 0–14 pH range with a sensitivity of 1 pH unit. Intermediate range indicators provide a sensitivity of 0.3–0.5 pH units and narrow range strips provide a sensitivity of 0.2–0.3 pH units.
MilliporeSigma # 1.09531.0001 replaces and is identical to MilliporeSigma # 9586-1.
MilliporeSigma # 1.09531.0007 replaces and is identical to MilliporeSigma # 9586-3.
MilliporeSigma # 1.09532.0001 replaces and is identical to MilliporeSigma # 9587-1.
MilliporeSigma # 1.09533.0001 replaces and is identical to MilliporeSigma # 9588-1.
MilliporeSigma # 1.09533.0007 replaces and is identical to MilliporeSigma # 9588-3.
MilliporeSigma # 1.09535.0001 replaces and is identical to MilliporeSigma # 9590-1.
MilliporeSigma # 1.09535.0007 replaces and is identical to MilliporeSigma # 9590-3.
MilliporeSigma # 1.09540.0001 replaces and is identical to MilliporeSigma # 9580-1.
MilliporeSigma # 1.09541.0001 replaces and is identical to MilliporeSigma # 9581-1.
MilliporeSigma # 1.09542.0001 replaces and is identical to MilliporeSigma # 9582-1.
MilliporeSigma # 1.09542.0007 replaces and is identical to MilliporeSigma # 9582-3.
MilliporeSigma # 1.09543.0001 replaces and is identical to MilliporeSigma # 9583-1.
MilliporeSigma # 1.09543.0007 replaces and is identical to MilliporeSigma # 9583-3.
MilliporeSigma # 1.09545.0001 replaces and is identical to MilliporeSigma # 9585-1.
MilliporeSigma # 1.09584.0001 replaces and is identical to MilliporeSigma # 9578-1.
 : Analyze your nitrate test strips with the reference card and with the free MQuant® StripScan app. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.The StripScan app allows the customer to analyze the MQuant® test strips with your smartphone for fast, convenient, and precise results which can be easily graphically presented and shared.
Specifications
  • Description:
    Acilit® pH-Indicator strips, non-bleeding, intermediate range
  • pH range:
    0 - 6
  • pH graduation:
    0-0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-
    4.5-5.0-5.5-6.0
  • Resolution:
    0.5
  • No. of strips:
    600