Phenol red solution pH-indicator
Dropping bottle, marked, 100 ml
Catalog # CA97026-640
Supplier: Hach
: 21132
Jump to:
Product Details & Documents
pH Indicator Solution
- Color range: pH 6.5-8.5
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Dropping bottle, marked
- Size:100 ml



