Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Hach

pH Buffer Solutions, Hach

±0.02, 25 °C, 500 mL, 4.01, Red
Catalog # CA97021-520
Supplier:   Hach
Supplier #:  2283449
pH Buffer Solutions, Hach
Catalog # CA97021-520
Supplier:  Hach
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 500 mL, 4.01, Red

CAS Number:  
Price
$57.40
Log in for availability
Each (500ml)
Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Specifications

Specifications

  • Color:
    Red
  • Size:
    500 mL
  • Reference temperature:
    25 °C
  • pH:
    4.01
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.