Trays, Rectangular, Polypropylene, Dynalon
18"L x 18"L x 4"H
Catalog # 30621-022
Supplier: Dynalon
Supplier #: 107324
Jump to:
Product Details & Documents
Heavy trays fabricated with 3/16" thick autoclavable Polypropylene
All welded construction. Trays can be customized with spigots and covers which are fabricated to order.
Ordering information: Additional sizes and materials are available upon request.
Specifications
- Interior dimensions:18"L x 18"L x 4"H