100-Well Storage Racks, Electron Microscopy Sciences
Product Details & Documents
Polypropylene rack features durable three point hinges and clasp for secure lid closure.
- Holds 1.5 to 2.0 ml microtubes and cryovials
- Rack measures 5.625×5.875×2.25"
User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base and imprinted and raised coordinates on the box.
Specifications
- Color:Blue
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)