Order Entry
Canada
Orders LinkContactUsLinkComponent
Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific

Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific

500 mL
Catalog # CAAAJ63974-AP
Supplier #:  J63974-AP
Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific
Catalog # CAAAJ63974-AP
CAS Number:  
Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific

500 mL

CAS Number:  
Price
$276.48
Log in for availability
Each (500ml)
Minimum order quantity is 1
Product Details & Documents

Liquid

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Documents

Specifications

Specifications

  • Size:
    500 mL
  • Cat. No.:
    CAAAJ63974-AP
  • MDL No.:
    MFCD00003527

Specifications