MOPS, 0.5 M Buffer Solutions, Thermo Scientific
MOPS buffer solution, 0.5 M, 250 mL, 6.5
Supplier: Thermo Scientific Chemicals
Supplier #: J63369-AK
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ63369-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183
Documents
Specifications
Specifications
- Concentration:0.5 M
- Description:MOPS buffer solution
- Size:250 mL
- pH:6.5
- Cat. No.:CAAAJ63369-AK
- Odour/scent:Odorless
- Storage temperature:2°C to 8°C
- MDL No.:MFCD00006183
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



