Order Entry
Canada
Orders LinkContactUsLinkComponent
 

MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS buffer solution, 0.5 M, 100 mL, 6.5
Catalog # CAAAJ63369-AE
 :  J63369-AE
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ63369-AE
CAS Number:  
MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS buffer solution, 0.5 M, 100 mL, 6.5

CAS Number:  
Price
$120.59
Log in for availability
Each (100ml)
 
Product Details & Documents

Liquid

CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183

Specifications

Specifications

  • Concentration:
    0.5 M
  • Description:
    MOPS buffer solution
  • Size:
    100 mL
  • pH:
    6.5
  • Cat. No.:
    CAAAJ63369-AE
  • Odour/scent:
    Odorless
  • Storage temperature:
    2°C to 8°C
  • MDL No.:
    MFCD00006183

Specifications