RIPA buffer (2X), Thermo Scientific
125ml
Supplier: Thermo Scientific Chemicals
Supplier #: J63324-EQE
RIPA buffer (2X), Thermo Scientific
Catalog # CAAAJ63324-EQE
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484
Documents
Specifications
Specifications
- Size:125ml
- Cat. No.:CAAAJ63324-EQE
- MDL No.:MFCD02100484
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



