Order Entry
Canada
Orders LinkContactUsLinkComponent
RIPA buffer (2X), Thermo Scientific

RIPA buffer (2X), Thermo Scientific

250 mL
Catalog # CAAAJ63324-AK
Supplier #:  J63324-AK
RIPA buffer (2X), Thermo Scientific
Catalog # CAAAJ63324-AK
Supplier:  
CAS Number:  
RIPA buffer (2X), Thermo Scientific

250 mL

CAS Number:  
Discontinued
Product Details & Documents

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484

Specifications

Specifications

  • Size:
    250 mL
  • Cat. No.:
    CAAAJ63324-AK
  • MDL No.:
    MFCD02100484

Specifications