Order Entry
Canada
Orders LinkContactUsLinkComponent
 

HEPES protein extraction buffer (5X)

250 mL
  CAAAJ63297-AK
 :  J63297-AK
HEPES protein extraction buffer (5X)
  CAAAJ63297-AK
 :  
HEPES protein extraction buffer (5X)

250 mL

 :  
 
78,33 $
   
Each (250ml)
 
 

Liquid

Physical form: liquid
Appearance: colorless

 

 

  • Size:
    250 mL