Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Thrombin Buffer, pH 8.0, Thermo Scientific

500 mL
  CAAAJ63240-AP
 :  J63240-AP
Thrombin Buffer, pH 8.0, Thermo Scientific
  CAAAJ63240-AP
 :  
Thrombin Buffer, pH 8.0, Thermo Scientific

500 mL

 :  
 
$185.94
   
Each (500ml)
 
 

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless

 

 

  • Size:
    500 mL