Order Entry
Canada
Orders LinkContactUsLinkComponent
Thrombin Buffer, pH 8.0, Thermo Scientific

Thrombin Buffer, pH 8.0, Thermo Scientific

250 mL
Catalog # CAAAJ63240-AK
Supplier #:  J63240-AK
Thrombin Buffer, pH 8.0, Thermo Scientific
Catalog # CAAAJ63240-AK
CAS Number:  
Thrombin Buffer, pH 8.0, Thermo Scientific

250 mL

CAS Number:  
Price
$65.32
Log in for availability
Each (250ml)
Minimum order quantity is 1
Product Details & Documents

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless

Specifications

Specifications

  • Size:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.