Order Entry
Canada
Orders LinkContactUsLinkComponent
Alkaline running buffer solution, Thermo Scientific

Alkaline running buffer solution, Thermo Scientific

500 mL
Catalog # CAAAJ63170-AP
Supplier #:  J63170-AP
Alkaline running buffer solution, Thermo Scientific
Catalog # CAAAJ63170-AP
CAS Number:  
Alkaline running buffer solution, Thermo Scientific

500 mL

CAS Number:  
Price
$102.26
Log in for availability
Each (500ml)
Minimum order quantity is 1
Product Details & Documents
Specifications

Specifications

  • Size:
    500 mL
  • Cat. No.:
    CAAAJ63170-AP
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.