Order Entry
Canada
Orders LinkContactUsLinkComponent
RNA sample loading buffer, Thermo Scientific

RNA sample loading buffer, Thermo Scientific

1 mL
Catalog # CAAAJ62832-XF
Supplier #:  J62832-XF
RNA sample loading buffer, Thermo Scientific
Catalog # CAAAJ62832-XF
CAS Number:  
RNA sample loading buffer, Thermo Scientific

1 mL

CAS Number:  
Price
$155.32
Log in for availability
Each (1ml)
Minimum order quantity is 1
Product Details & Documents

Liquid

Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00213041

Documents

Specifications

Specifications

  • Size:
    1 mL
  • Cat. No.:
    CAAAJ62832-XF
  • MDL No.:
    MFCD00213041

Specifications