Zymogram sample buffer, pH 6.6, Thermo Scientific
50 mL
Zymogram sample buffer, pH 6.6, Thermo Scientific
Catalog # CAAAJ62774-AD
CAS Number:
Jump to:
Discontinued
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Blue
Documents
Specifications
Specifications
- Size:50 mL



