Order Entry
Canada
Orders LinkContactUsLinkComponent
L-thyroxine 98%

L-thyroxine 98%

5 g
Catalog # CAAAJ62606-06
Supplier #:  J62606-06
L-thyroxine 98%
Catalog # CAAAJ62606-06
CAS Number:  51-48-9
L-thyroxine 98%

5 g

CAS Number:  51-48-9
Price
$658.29
Log in for availability
Each (5g)
Minimum order quantity is 1
Product Details & Documents

Crystalline powder

Purity: 98% CAS No.: 51-48-9 Molecular Formula: C15H11I4NO4 Formula Weight: 776.87 Physical Form: Crystalline powder Sensitivity: Light Sensitive Appearance: White to pale brown Odor: Odorless Melting Point: 235-236° Solubility: Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml Beilstein Reference: 2228515 MDL No.: MFCD00002595

Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • MDL Number:MFCD00002595
  • Molecular Weight:776.874 g/mol
  • Merck Index:14,9415
  • Melting Pt:235…236 °C
  • Storage Temperature:Ambient
  • EINECS:200-101-1
Specifications

Specifications

  • Size:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.